Learn More
4-Chloro-N-cyclohexylbutyramide, 97%, Thermo Scientific™
Product Code. 15496148
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496148 250 mg 250mg
15486148 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15496148 Supplier Thermo Scientific Alfa Aesar Supplier No. H59752.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Este artículo no se puede devolver. Vea la política de devoluciones
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 69601-43-0
Molecular Formula C10H18ClNO
Molecular Weight (g/mol) 203.71
MDL Number MFCD02729224
InChI Key KYKIYEJLXULPLV-UHFFFAOYSA-N
Synonym historical, 4-chloro-n-cyclohexylbutyramide, n-4-chlorobutyryl cyclohexylamine, n-gamma-chlorobutyrylcyclohexylamine, butanamide, 4-chloro-n-cyclohexyl
PubChem CID 4103470
IUPAC Name 4-chloro-N-cyclohexylbutanamide
SMILES C1CCC(CC1)NC(=O)CCCCl
Especificaciones
Quantity 250 mg
Formula Weight 203.71
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-cyclohexylbutyramide
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.