accessibility menu, dialog, popup

UserName

H3COHN(S)NH2OHO

CAS RN 692-04-6

IUPAC Name: 2-amino-6-acetamidohexanoic acid
Synonyms: acetyllysine
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: DTERQYGMUDWYAZ-UHFFFAOYNA-N
SMILES: CC(=O)NCCCCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)