missing translation for 'onlineSavingsMsg'
Learn More

N-ε-Acetyl-L-Lysine, MP Biomedicals™

Product Code. 11453041 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453041 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453041 Supplier MP Biomedicals Supplier No. 02150235.2
  • ONLINE EXCLUSIVE PRICE
    45.20 EUR
    43.05 EUR / 250mg
    Save 2.15 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used together with other lysine analogues. N-ε-Acetyl-L-Lysine, MP Biomedicals (L-AcK) is an R-chain N-acetylated α amino acid used to differentiate and characterize various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

TRUSTED_SUSTAINABILITY

Specifications

CAS 692-04-6
Melting Point 247.1°C to 247.3°C (literature)
Molecular Formula C8H16N2O3
MDL Number MFCD00002639
Quantity 250 mg
Synonym n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine
Solubility Information Soluble in 80% AcOH (5mg/mL, clear and colorless solution) and water.
InChI Key DTERQYGMUDWYAZ-ZETCQYMHSA-N
SMILES CC(=O)NCCCC[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-6-acetamidohexanoic acid
Molecular Weight (g/mol) 188.23
PubChem CID 92832
ChEBI CHEBI:17752
Formula Weight 188.26
Percent Purity ≥99%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.