missing translation for 'onlineSavingsMsg'
Learn More

N-Epsilon-acetyl-l-lysine, TRC

Product Code. 30135455
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135455 5 g 5g
30135375 10 g 10g
30012401 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135455 Supplier Toronto Research Chemicals Supplier No. TRCE5887255G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 692-04-6
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.22
Synonym N6-Acetyl-L-lysine, L-ε-N-Acetyllysine, N6-Acetyllysine, NSC 102777, Nε-Acetyl-L-lysine, Nε-Acetyllysine, ε-Acetyl-L-lysine, ε-N-Acetyl-L-lysine, ε-N-Acetyllysine, ω-N-Acetyl-L-lysine
SMILES CC(NCCCC[C@H](N)C(O)=O)=O

Specifications

InChI Formula InChI=1S/C8H16N2O3/c1-6(11)10-5-3-2-4-7(9)8(12)13/h7H,2-5,9H2,1H3,(H,10,11)(H,12,13)/t7-/m0/s1
Quantity 5 g
Formula Weight 188.1161
Shipping Condition Room Temperature
Chemical Name or Material N-Epsilon-acetyl-l-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.