CAS RN 68377-33-3
IUPAC Name:
1-(4-methoxyphenyl)-1H-1,2,4-triazole
Synonyms:
1-(4-methoxyphenyl)-1,2,4-triazole
Molecular Weight (g/mol):
175.19
Molecular Formula:
C9H9N3O
InChi Key:
HXCCOAUMJBFQCC-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)N1C=NC=N1
1
–
1
of
1
results
1
Alfa Aesar™ 1-(4-Methoxyphenyl)-1H-1,2,4-triazole, 99%
CAS: 68377-33-3 Molecular Formula: C9H9N3O Molecular Weight (g/mol): 175.191 MDL Number: MFCD12407079 InChI Key: HXCCOAUMJBFQCC-UHFFFAOYSA-N Synonym: 1-4-methoxyphenyl-1h-1,2,4-triazole,1-4-methoxyphenyl-1,2,4-triazole,1-4-methoxyphenyl-1h-1,2,4 triazole,4-n-methoxyphenyl-1,2,4-triazole,1-4-methoxy-phenyl-1h-1,2,4 triazole PubChem CID: 11084390 IUPAC Name: 1-(4-methoxyphenyl)-1,2,4-triazole SMILES: COC1=CC=C(C=C1)N2C=NC=N2