missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Methoxyphenyl)-1H-1,2,4-triazole, 99%

Product Code. 15444017
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444017 25 g 25g
15434017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444017 Supplier Alfa Aesar™ Supplier No. H30962.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68377-33-3
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD12407079
InChI Key HXCCOAUMJBFQCC-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl-1h-1,2,4-triazole, 1-4-methoxyphenyl-1,2,4-triazole, 1-4-methoxyphenyl-1h-1,2,4 triazole, 4-n-methoxyphenyl-1,2,4-triazole, 1-4-methoxy-phenyl-1h-1,2,4 triazole
PubChem CID 11084390
IUPAC Name 1-(4-methoxyphenyl)-1,2,4-triazole
SMILES COC1=CC=C(C=C1)N2C=NC=N2

Specifications

Melting Point 96°C to 98°C
Quantity 25 g
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material 1-(4-Methoxyphenyl)-1H-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.