accessibility menu, dialog, popup

UserName

CAS RN 661-71-2

IUPAC Name: 1-chloro-1,2,2-trifluorocyclobutane
Synonyms: 1-chloro-1,2,2-trifluorocyclobutane
Molecular Weight (g/mol): 144.52
Molecular Formula: C4H4ClF3
InChi Key: ZZQYDYODFHABLC-UHFFFAOYNA-N
SMILES: FC1(F)CCC1(F)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results