missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-1,2,2-trifluorocyclobutane, 97%, Thermo Scientific™

Product Code. 11359165
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359165 25 g 25g
11349165 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359165 Supplier Thermo Scientific Alfa Aesar Supplier No. L16436.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-71-2
Molecular Formula C4H4ClF3
Molecular Weight (g/mol) 144.521
MDL Number MFCD00042139
InChI Key ZZQYDYODFHABLC-UHFFFAOYSA-N
PubChem CID 79091
IUPAC Name 1-chloro-1,2,2-trifluorocyclobutane
SMILES C1CC(C1(F)F)(F)Cl

Specifications

Density 1.34
Boiling Point 81°C
Refractive Index 1.357
Quantity 25 g
UN Number UN1993
Beilstein 1902628
Synonym cyclobutane, 1-chloro-1,2,2-trifluoro, 1-ctfcb, d0p0zb, cyclobutane,1-chloro-1,2,2-trifluoro
Formula Weight 144.52
Percent Purity 97%
Chemical Name or Material 1-Chloro-1,2,2-trifluorocyclobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.