accessibility menu, dialog, popup

UserName

OHN

CAS RN 65195-20-2

IUPAC Name: 2-(piperidin-1-yl)phenol
Synonyms: 2-(piperidin-1-yl)phenol
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: OXELIFPFCCGAJX-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results