missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperidinyl)phenol, 98%

Product Code. 11341997
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341997 5 g 5g
11331997 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341997 Supplier Thermo Scientific Alfa Aesar Supplier No. L07630.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 65195-20-2 | C11H15NO | 177.25 g/mol

2-(1-Piperidinyl)phenol is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(1-Piperidinyl)phenol is used as primary and secondary intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65195-20-2
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00051687
InChI Key OXELIFPFCCGAJX-UHFFFAOYSA-N
Synonym 2-piperidin-1-yl phenol, 2-piperidinophenol, 2-1-piperidino phenol, 2-1-piperidinyl phenol, 2-piperidylphenol, 2-hydroxyphenyl piperidine, 1-2-hydroxyphenyl piperidine
PubChem CID 3262250
SMILES OC1=CC=CC=C1N1CCCCC1

Specifications

Melting Point 73°C to 76°C
Quantity 5 g
Beilstein 151777
IUPAC Name 2-piperidin-1-ylphenol
Formula Weight 177.24
Percent Purity 98%
Chemical Name or Material 2-(1-Piperidinyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.