accessibility menu, dialog, popup

UserName

CAS RN 65-28-1

CH3NNHNHOCH3SOHOO

CAS RN 65-28-1

IUPAC Name: 3-{[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino}phenol; methanesulfonic acid
Synonyms: methanesulfonic acid; phentolamine
Molecular Weight (g/mol): 377.46
Molecular Formula: C18H23N3O4S
InChi Key: OGIYDFVHFQEFKQ-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CC1=CC=C(C=C1)N(CC1=NCCN1)C1=CC=CC(O)=C1
Quantity 
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results