accessibility menu, dialog, popup

UserName

H3CONSHNHBr

CAS RN 63208-82-2

IUPAC Name: 2-(2-imino-2,3,4,5,6,7-hexahydro-1,3-benzothiazol-3-yl)-1-(4-methylphenyl)ethan-1-one hydrobromide
Synonyms: pifithrin
Molecular Weight (g/mol): 367.31
Molecular Formula: C16H19BrN2OS
InChi Key: HAGVCKULCLQGRF-UHFFFAOYSA-N
SMILES: Br.CC1=CC=C(C=C1)C(=O)CN1C(=N)SC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results