missing translation for 'onlineSavingsMsg'
Learn More

Pifithrin-α hydrobromide, MedChemExpress

Product Code. 16499137
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499137 5 mg 5mg
16483476 25 mg 25mg
16462657 50 mg 50mg
16446568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499137 Supplier Medchem Express Supplier No. HY15484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pifithrin-α hydrobromide is a p53 inhibitor which blocks its transcriptional activity and prevents cells from apoptosis. Pifithrin-α hydrobromide is also an aryl hydrocarbon receptor (AhR) agonist.

Pifithrin-α hydrobromide is a p53 inhibitor which blocks its transcriptional activity and prevents cells from apoptosis. Pifithrin-α hydrobromide is also an aryl hydrocarbon receptor (AhR) agonist.

Specifications

Chemical Name or Material Pifithrin-α hydrobromide
Physical Form Solid
CAS 63208-82-2
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H19BrN2OS
For Use With (Application) Cancer-programmed cell death
Synonym Pifithrin hydrobromide PFTα hydrobromide
Solubility Information DMSO : ≥ 50 mg/mL (136.13 mM) ∣H2O : 1.25 mg/mL (3.40 mM; Need ultrasonic)
SMILES N=C1SC2=C(CCCC2)N1CC(C3=CC=C(C)C=C3)=O.[H]Br
Molecular Weight (g/mol) 367.3
Formula Weight 367.3
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.