accessibility menu, dialog, popup

UserName

NHF

CAS RN 625437-46-9

IUPAC Name: N-[(4-fluorophenyl)methyl]cyclopropanamine
Synonyms: N-[(4-fluorophenyl)methyl]cyclopropanamine
Molecular Weight (g/mol): 165.21
Molecular Formula: C10H12FN
InChi Key: YKNFXOCMGURJMK-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results