missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclopropyl-4-fluorobenzylamine, 97%, Thermo Scientific™

Product Code. 15432648
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432648 250 mg 250mg
15422648 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432648 Supplier Thermo Scientific Alfa Aesar Supplier No. H56206.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625437-46-9
Molecular Formula C10H12FN
Molecular Weight (g/mol) 165.21
MDL Number MFCD07330007
InChI Key YKNFXOCMGURJMK-UHFFFAOYSA-N
Synonym n-4-fluorobenzyl cyclopropanamine, cyclopropyl-4-fluoro-benzyl-amine, n-4-fluorophenyl methyl cyclopropanamine, cyclopropyl-4-fluorobenzyl amine, benzenemethanamine, n-cyclopropyl-4-fluoro, n-cyclopropyl-p-fluorobenzylamine, acmc-209n4r, n-cyclopropyl-4-fluorobenzylamine, n-4-fluorobenzyl cyclopropanamine #, n-4-fluorophenyl methyl cyclopropylamine
PubChem CID 573031
IUPAC Name N-[(4-fluorophenyl)methyl]cyclopropanamine
SMILES FC1=CC=C(CNC2CC2)C=C1

Specifications

Quantity 250 mg
UN Number UN2735
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material N-Cyclopropyl-4-fluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.