accessibility menu, dialog, popup

UserName

CAS RN 616-91-1

IUPAC Name: 2-acetamido-3-sulfanylpropanoic acid
Synonyms: acetylcysteine
Molecular Weight (g/mol): 163.19
Molecular Formula: C5H9NO3S
InChi Key: PWKSKIMOESPYIA-UHFFFAOYNA-N
SMILES: CC(=O)NC(CS)C(O)=O
Molecular Weight (g/mol) 
  • (15)
  • (12)
  • (3)
Quantity 
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (4)
Grade 
  • (4)
  • (9)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (9)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (15)
  • (12)
  • (3)
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (4)
  • (4)
  • (9)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)