missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-cysteine (Crystalline), MP Biomedicals

Product Code. 11495012 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Unit Size:
100g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495012 100 g 100g
11465012 5 g 5g
11475012 25 g 25g
11485012 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11495012 Supplier MP Biomedicals Supplier No. 0219460380
  • ONLINE EXCLUSIVE PRICE
    217.00 EUR
    205.80 EUR / 100g
    Save 11.20 EUR
    5% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-91-1 | C5H9NO3S | 163.191 g/mol

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Color White
Alpha Vector Acetyl-L-cysteine
Quantity 100 g
Formula Weight 163.2
Percent Purity ≥99.9%
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Assay ≥96%
Chemical Name or Material N-Acetyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.