accessibility menu, dialog, popup

UserName

FFFNClClO

CAS RN 61213-25-0

IUPAC Name: 3-chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]pyrrolidin-2-one
Synonyms: flurochloridone epa pesticide chemical code 128401 racer raiser caswell no. 721B C12H10cl2F3NO fluorochloridone
Molecular Weight (g/mol): 312.11
Molecular Formula: C12H10Cl2F3NO
InChi Key: OQZCSNDVOWYALR-UHFFFAOYNA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)N1CC(CCl)C(Cl)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Concentration 
  • (4)
Solvent or Matrix 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)