Learn More
Flurochloridon, TRC
Product Code. 30135066
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135066 1 g 1g
30041223 100 mg 100mg
18090070 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135066 Supplier Toronto Research Chemicals Supplier No. TRCF5990331G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61213-25-0
Molecular Formula C12 H10 Cl2 F3 N O
Molecular Weight (g/mol) 312.12
Synonym 3-Chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]-2-pyrrolidinone, Fluorochloridone, Flurochloridone, R 40244, Racer, Racer 25, Racer 250 EC, Rainbow, Twin Gold Pack, 2-Pyrrolidinone, 3-chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]-
IUPAC Name 3-chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]pyrrolidin-2-one
SMILES FC(F)(F)c1cccc(c1)N2CC(CCl)C(Cl)C2=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C12H10Cl2F3NO/c13-5-7-6-18(11(19)10(7)14)9-3-1-2-8(4-9)12(15,16)17/h1-4,7,10H,5-6H2
Formula Weight 311.0092
Chemical Name or Material Flurochloridone