missing translation for 'onlineSavingsMsg'
Learn More

Flurochloridone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316833
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316833 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316833 Supplier Dr. Ehrenstorfer Supplier No. DREXA13847500AL
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flurochloridone
CAS 61213-25-0
Analyte or Component Names Flurochloridone
Shipping Condition Room Temperature
Molecular Formula C12 H10 Cl2 F3 N O
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym 3-Chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]-2-pyrrolidinone, Fluorochloridone, Flurochloridone, R 40244, Racer, Racer 25, Racer 250 EC, Rainbow, Twin Gold Pack, 2-Pyrrolidinone, 3-chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]-
InChI Formula InChI=1S/C12H10Cl2F3NO/c13-5-7-6-18(11(19)10(7)14)9-3-1-2-8(4-9)12(15,16)17/h1-4,7,10H,5-6H2
Solution Type Single Solution
SMILES FC(F)(F)c1cccc(c1)N2CC(CCl)C(Cl)C2=O
IUPAC Name 3-chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]pyrrolidin-2-one
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 312.12
Quantity 1 mL
Formula Weight 311.0092 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.