accessibility menu, dialog, popup

UserName

H3COOCH3H3COOH3COOH3COOCH3

CAS RN 6077-72-1

IUPAC Name: 1,1,2,2-tetramethyl 3-(2-methylpropyl)cyclopropane-1,1,2,2-tetracarboxylate
Synonyms: 1,1,2,2-tetramethyl 3-(2-methylpropyl)cyclopropane-1,1,2,2-tetracarboxylate
Molecular Weight (g/mol): 330.33
Molecular Formula: C15H22O8
InChi Key: CFXMPMLQBIHACL-UHFFFAOYSA-N
SMILES: COC(=O)C1(C(CC(C)C)C1(C(=O)OC)C(=O)OC)C(=O)OC
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)