missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopropanemethanol, 98%, mixture of cis and trans, Thermo Scientific™

Product Code. 10763021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763021 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10763021 Supplier Thermo Scientific Acros Supplier No. 214940010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6077-72-1
Molecular Formula C15H22O8
Molecular Weight (g/mol) 330.33
MDL Number MFCD00009683
InChI Key CFXMPMLQBIHACL-UHFFFAOYSA-N
Synonym 2-methylcyclopropyl methanol, 2-methylcyclopropanemethanol, cyclopropanemethanol, 2-methyl, 2-methylcyclopropanemethanol, mixture of cis and trans, acmc-20a6ci, 2-methyl-cyclopropane-methanol, cyclopropanemethanol,2-methyl, 2-methylcyclopropyl methan-1-ol, 2-methylcyclopropanemethanol, cis + trans
PubChem CID 138655
SMILES COC(=O)C1(C(CC(C)C)C1(C(=O)OC)C(=O)OC)C(=O)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8700g/mL
Boiling Point 133.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C3H4CH2OH
Refractive Index 1.4280 to 1.4330
Quantity 1 g
Specific Gravity 0.87
IUPAC Name (2-methylcyclopropyl)methanol
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclopropanemethanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylcyclopropanemethanol, mixture of cis and trans
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.