missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopropanemethanol, cis + trans, 98%, Thermo Scientific™

Product Code. 11312439
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312439 10 g 10g
11302439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312439 Supplier Thermo Scientific Alfa Aesar Supplier No. H27337.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the radical clock investigation of microsomal cytochrome P-450 hydroxylation of hydrocarbons.

Solubility
Slightly soluble in water.

Notes
Store away from halogens and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away and protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6077-72-1
Molecular Formula C15H22O8
Molecular Weight (g/mol) 330.33
MDL Number MFCD00009683
InChI Key CFXMPMLQBIHACL-UHFFFAOYSA-N
Synonym 2-methylcyclopropyl methanol, 2-methylcyclopropanemethanol, cyclopropanemethanol, 2-methyl, 2-methylcyclopropanemethanol, mixture of cis and trans, acmc-20a6ci, 2-methyl-cyclopropane-methanol, cyclopropanemethanol,2-methyl, 2-methylcyclopropyl methan-1-ol, 2-methylcyclopropanemethanol, cis + trans
PubChem CID 138655
IUPAC Name (2-methylcyclopropyl)methanol
SMILES COC(=O)C1(C(CC(C)C)C1(C(=O)OC)C(=O)OC)C(=O)OC

Specifications

Density 0.87
Boiling Point 133°C
Flash Point 39°C (102°F)
Odor Alcohol-like
Refractive Index 1.432
Quantity 10 g
UN Number UN1987
Solubility Information Slightly soluble in water.
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material 2-Methylcyclopropanemethanol, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.