accessibility menu, dialog, popup

UserName

H2OCH3NONCH3NHNO

CAS RN 5967-84-0

IUPAC Name: 1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione hydrate
Synonyms: theophylline hydrate
Molecular Weight (g/mol): 198.18
Molecular Formula: C7H10N4O3
InChi Key: INQSMEFCAIHTJG-UHFFFAOYSA-N
SMILES: O.CN1C2=C(NC=N2)C(=O)N(C)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)