Learn More
Theophylline monohydrate, 99%, Thermo Scientific™
24.55 EUR - 287.00 EUR
Chemical Identifiers
| CAS | 5967-84-0 |
|---|---|
| Molecular Formula | C7H10N4O3 |
| Molecular Weight (g/mol) | 198.182 |
| MDL Number | MFCD00151659 |
| InChI Key | INQSMEFCAIHTJG-UHFFFAOYSA-N |
| Synonym | theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 |
| PubChem CID | 91268 |
| IUPAC Name | 1,3-dimethyl-7H-purine-2,6-dione;hydrate |
| SMILES | CN1C2=C(C(=O)N(C1=O)C)NC=N2.O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water (8 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| C7H10N4O3 | |
| MFCD00151659 | |
| theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 | |
| 1,3-dimethyl-7H-purine-2,6-dione;hydrate |
Specifications
| 5967-84-0 | |
| C7H10N4O3 | |
| UN2811 | |
| 14,9285 | |
| Soluble in water (8g/L at 20°C). | |
| CN1C2=C(C(=O)N(C1=O)C)NC=N2.O | |
| 198.182 | |
| 198.18 (180.16 Anhydrous) | |
| Theophylline monohydrate |
| Odorless | |
| MFCD00151659 | |
| 13463 | |
| theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 | |
| INQSMEFCAIHTJG-UHFFFAOYSA-N | |
| 1,3-dimethyl-7H-purine-2,6-dione;hydrate | |
| 91268 | |
| 99% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 200-385-7
missing translation for 'rtecsNumber' : XH3850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.