Learn More
Theophylline monohydrate, 99%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar A15997.30
| Quantity | 250g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water (8 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| C7H10N4O3 | |
| MFCD00151659 | |
| theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 | |
| 1,3-dimethyl-7H-purine-2,6-dione;hydrate |
Specifications
| 5967-84-0 | |
| C7H10N4O3 | |
| 250g | |
| 13463 | |
| theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 | |
| INQSMEFCAIHTJG-UHFFFAOYSA-N | |
| 1,3-dimethyl-7H-purine-2,6-dione;hydrate | |
| 91268 | |
| 99% |
| Odorless | |
| MFCD00151659 | |
| UN2811 | |
| 14,9285 | |
| Soluble in water (8g/L at 20°C). | |
| CN1C2=C(C(=O)N(C1=O)C)NC=N2.O | |
| 198.182 | |
| 198.18 (180.16 Anhydrous) | |
| Theophylline monohydrate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.