CAS RN 59-47-2
CAS RN 59-47-2
IUPAC Name:
3-(2-methylphenoxy)propane-1,2-diol
Synonyms:
mephenesin sanofi synthelabo otc brand of mephenesin mephenesinum décontractyl rhex 'hobeino' mephenesin (substance) o-kresol-glycerinaether mephelor cresoxydiol kade brand of mephenesin
Molecular Weight (g/mol):
182.22
Molecular Formula:
C10H14O3
InChi Key:
JWDYCNIAQWPBHD-UHFFFAOYNA-N
SMILES:
CC1=CC=CC=C1OCC(O)CO
1
–
3
of
3
results
1
Mephenesin, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Mephenesin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Mephenesin, MedChemExpress
MedChemExpress Mephenesin is an NMDA receptor antagonist, is a centrally acting muscle relaxant.