Learn More
Mephenesin, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Mephenesin
Specifications
Specifications
| Chemical Name or Material | Mephenesin |
| CAS | 59-47-2 |
| Analyte or Component Names | Mephenesin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H14 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mephenesin, 1,2-Propanediol, 3-(2-methylphenoxy)-, 1,2-Propanediol, 3-(o-tolyloxy)- (8CI), 3-(2-Methylphenoxy)-1,2-propanediol, (+/-)-Mephenesin, 1,2-Dihydroxy-3-(2-methylphenoxy)propane, 3-(2-Methylphenoxy)propane-1,2-diol, 3-(2-Tolyloxy)-1,2-propanediol, 3-(o-Methylphenoxy)-1,2-propanediol, 3-(o-Tolyloxy)-1,2-propanediol, 3-o-Toloxy-1,2-propanediol, Atensin, Avosyl, Avoxyl, BDH 312, Cresoxydiol, Cresoxypropanediol, Curythan, Daserd, Daserol, Decontractyl, Dioloxol, Glyceryl o-tolyl ether, Glykresin, Glyotol, Kinavosyl, Lissephen, Memphenesin, Mephedan, Mephelor, Mephenesine, Mephensin, Mepherol, Mephesin, Mephin, Mephson, Mervaldin, Myanesin, Myanol, Myasin, Mycocuran, Myodetensin, Myodetensine, Myolysin, Myopan, Myoserol, Myoten, Myoxane, NSC 25234, NSC 36140, NSC 50788, NSC 8134, Oranixon, Prolax, RP 3602, Relaxar, Relaxil, Renarcol, Rhex, Sansdolor, Sinan, Spartoloxyn, Spasmolyn, Stilalgin, Thoxidil, Tolansin, Tolax, Tolcil, Tolhart, Tolosate, Toloxyn, Tolserol, Tolseron, Tolsin, Tolulexin, Tolulox, Tolyspaz, Walconesin, o-Cresyl glycerol ether, o-Cresyl alpha-glyceryl ether |
| InChI Formula | InChI=1S/C10H14O3/c1-8-4-2-3-5-10(8)13-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3 |
| SMILES | Cc1ccccc1OCC(O)CO |
| IUPAC Name | 3-(2-methylphenoxy)propane-1,2-diol |
| Show More |