Learn More
Mephenesin, MedChemExpress
Product Code. 16452329
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 16452329
Brand: Medchem Express HYB1283
MedChemExpress Mephenesin is an NMDA receptor antagonist, is a centrally acting muscle relaxant.
Mephenesin is an NMDA receptor antagonist, is a centrally acting muscle relaxant.
Specifications
Chemical Name or Material Mephenesin
CAS 59-47-2
Color White
Purity Grade Notes Research
Molecular Formula C10H14O3
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (548.79 mM)
SMILES OCC(O)COC1=CC=CC=C1C
Molecular Weight (g/mol) 182.22
Formula Weight 182.22
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration