missing translation for 'onlineSavingsMsg'
Learn More

Mephenesin, LoGiCal

Product Code. 30459555
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30459555

Brand: LoGiCal LGCFOR0810.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mephenesin
CAS 59-47-2
Analyte or Component Names Mephenesin
Shipping Condition Room Temperature
Molecular Formula C10 H14 O3
Certifications/Compliance ISO 9001
Synonym Mephenesin, 1,2-Propanediol, 3-(2-methylphenoxy)-, 1,2-Propanediol, 3-(o-tolyloxy)- (8CI), 3-(2-Methylphenoxy)-1,2-propanediol, (+/-)-Mephenesin, 1,2-Dihydroxy-3-(2-methylphenoxy)propane, 3-(2-Methylphenoxy)propane-1,2-diol, 3-(2-Tolyloxy)-1,2-propanediol, 3-(o-Methylphenoxy)-1,2-propanediol, 3-(o-Tolyloxy)-1,2-propanediol, 3-o-Toloxy-1,2-propanediol, Atensin, Avosyl, Avoxyl, BDH 312, Cresoxydiol, Cresoxypropanediol, Curythan, Daserd, Daserol, Decontractyl, Dioloxol, Glyceryl o-tolyl ether, Glykresin, Glyotol, Kinavosyl, Lissephen, Memphenesin, Mephedan, Mephelor, Mephenesine, Mephensin, Mepherol, Mephesin, Mephin, Mephson, Mervaldin, Myanesin, Myanol, Myasin, Mycocuran, Myodetensin, Myodetensine, Myolysin, Myopan, Myoserol, Myoten, Myoxane, NSC 25234, NSC 36140, NSC 50788, NSC 8134, Oranixon, Prolax, RP 3602, Relaxar, Relaxil, Renarcol, Rhex, Sansdolor, Sinan, Spartoloxyn, Spasmolyn, Stilalgin, Thoxidil, Tolansin, Tolax, Tolcil, Tolhart, Tolosate, Toloxyn, Tolserol, Tolseron, Tolsin, Tolulexin, Tolulox, Tolyspaz, Walconesin, o-Cresyl glycerol ether, o-Cresyl alpha-glyceryl ether
InChI Formula InChI=1S/C10H14O3/c1-8-4-2-3-5-10(8)13-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3
Solution Type Neat
SMILES Cc1ccccc1OCC(O)CO
IUPAC Name 3-(2-methylphenoxy)propane-1,2-diol
Industry Type Clinical Testing
Molecular Weight (g/mol) 182.22
Quantity 10 mg
Formula Weight 182.0943 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.