accessibility menu, dialog, popup

UserName

CH3ONHCl

CAS RN 588-07-8

IUPAC Name: N-(3-chlorophenyl)acetamide
Synonyms: 3-chloroacetanilide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: MUUQHCOAOLLHIL-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results