Learn More
3-Chloroacetanilide, TRC
Product Code. 30137520
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137520 10 g 10g
30008555 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137520 Supplier Toronto Research Chemicals Supplier No. TRCC32537010G
 Restricted Item
Estimated Shipment: 25-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 588-07-8
Molecular Formula C8 H8 Cl N O
Molecular Weight (g/mol) 169.608
Synonym N-(3-Chlorophenyl)acetamide, NSC 39967, NSC 6116, m-Chloroacetanilide
IUPAC Name N-(3-chlorophenyl)acetamide
SMILES CC(=O)Nc1cccc(Cl)c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C8H8ClNO/c1-6(11)10-8-4-2-3-7(9)5-8/h2-5H,1H3,(H,10,11)
Formula Weight 169.029
Chemical Name or Material 3-Chloroacetanilide