Learn More
3'-Chloroacetanilide, 98+%, Thermo Scientific™
Product Code. 11482737
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482737 100 g 100g
11472737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11482737 Supplier Thermo Scientific Alfa Aesar Supplier No. A14921.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 588-07-8
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00016341
InChI Key MUUQHCOAOLLHIL-UHFFFAOYSA-N
Synonym 3'-chloroacetanilide, 3-chloroacetanilide, n-3-chlorophenyl acetamide, m-chloroacetanilide, acetamide, n-3-chlorophenyl, acetanilide, 3'-chloro, meta-chloroacetanilide, unii-74h09hu61j, acetic acid, amide, n 3-chlorophenyl, m-chloroacetoanilide
PubChem CID 11493
IUPAC Name N-(3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)Cl
Specifications
Melting Point 75°C to 79°C
Quantity 100 g
Beilstein 2208105
Merck Index 14,2111
Formula Weight 169.61
Percent Purity ≥98%
Chemical Name or Material 3'-Chloroacetanilide