CAS RN 57818-37-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methyl-5-pentyl-2-furanundecanoic Acid, TRC
CAS: 57818-37-8 Molecular Formula: C21H36O3 Molecular Weight (g/mol): 336.51 Synonym: 12,15-Epoxy-13-methyleicosa-12,14-dienoic Acid,F6 acid SMILES: CCCCCc1oc(CCCCCCCCCCC(=O)O)c(C)c1