missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-pentyl-2-furanundecanoic Acid, TRC

Product Code. 30138148
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138148 50 mg 50mg
30004923 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138148 Supplier Toronto Research Chemicals Supplier No. TRCM32038550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57818-37-8
Molecular Formula C21H36O3
Molecular Weight (g/mol) 336.51
Synonym 12,15-Epoxy-13-methyleicosa-12,14-dienoic Acid, F6 acid
SMILES CCCCCc1oc(CCCCCCCCCCC(=O)O)c(C)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H36O3/c1-3-4-11-14-19-17-18(2)20(24-19)15-12-9-7-5-6-8-10-13-16-21(22)23/h17H,3-16H2,1-2H3,(H,22,23)
Formula Weight 336.2664
Percent Purity >95%
Chemical Name or Material 3-Methyl-5-pentyl-2-furanundecanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.