CAS RN 57817-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N6,O2'-Dimethyladenosine, TRC
CAS: 57817-83-1 Molecular Formula: C12H17N5O4 Molecular Weight (g/mol): 295.29 Synonym: N-Methyl-2'-O-methyladenosine,2'-O-Methyl-6-methyladenosine,6-Methyl-2'-O-methyladenosine,N6,2'-O-Dimethyladenosine,N6,O2'-Dimethyladenosine,N6-Methyl-2'-O-methyladenosine; SMILES: O[C@H]1[C@@H](OC)[C@H](N2C(N=CN=C3NC)=C3N=C2)O[C@@H]1CO