missing translation for 'onlineSavingsMsg'
Learn More

N6,O2'-Dimethyladenosine, TRC

Product Code. 30135614
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135614 50 mg 50mg
30133279 25 mg 25mg
17957544 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135614 Supplier Toronto Research Chemicals Supplier No. TRCD44741550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57817-83-1
Molecular Formula C12H17N5O4
Molecular Weight (g/mol) 295.29
Synonym N-Methyl-2'-O-methyladenosine, 2'-O-Methyl-6-methyladenosine, 6-Methyl-2'-O-methyladenosine, N6,2'-O-Dimethyladenosine, N6,O2'-Dimethyladenosine, N6-Methyl-2'-O-methyladenosine;
SMILES O[C@H]1[C@@H](OC)[C@H](N2C(N=CN=C3NC)=C3N=C2)O[C@@H]1CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H17N5O4/c1-13-10-7-11(15-4-14-10)17(5-16-7)12-9(20-2)8(19)6(3-18)21-12/h4-6,8-9,12,18-19H,3H2,1-2H3,(H,13,14,15)/t6-,8-,9-,12-/m1/s1
Formula Weight 295.1281
Percent Purity >95%
Chemical Name or Material N6,O2'-Dimethyladenosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.