CAS RN 56341-08-3
CAS RN 56341-08-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Mabuterol, TRC
CAS: 56341-08-3 Molecular Formula: C13 H18 Cl F3 N2 O Molecular Weight (g/mol): 310.74 Synonym: Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)-,4-Amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)benzenemethanol,(±)-Mabuterol,Ambuterol,Mabuterol,PB 868Cl IUPAC Name: 1-[4-amino-3-chloro-5-(trifluoromethyl)phenyl]-2-(tert-butylamino)ethanol SMILES: CC(C)(C)NCC(O)c1cc(Cl)c(N)c(c1)C(F)(F)F
2
Mabuterol 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality