missing translation for 'onlineSavingsMsg'
Learn More

Mabuterol 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313389
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313389 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313389 Supplier Dr. Ehrenstorfer Supplier No. DREA14659950AL1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mabuterol
CAS 56341-08-3
Analyte or Component Names Mabuterol
Shipping Condition +20°C
Molecular Formula C13 H18 Cl F3 N2 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)-, 4-Amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)benzenemethanol, (±)-Mabuterol, Ambuterol, PB 868Cl
InChI Formula InChI=1S/C13H18ClF3N2O/c1-12(2,3)19-6-10(20)7-4-8(13(15,16)17)11(18)9(14)5-7/h4-5,10,19-20H,6,18H2,1-3H3
SMILES CC(C)(C)NCC(O)c1cc(Cl)c(N)c(c1)C(F)(F)F
IUPAC Name 1-[4-amino-3-chloro-5-(trifluoromethyl)phenyl]-2-(tert-butylamino)ethanol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 310.74
Quantity 1 mL
Formula Weight 310.106 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.