missing translation for 'onlineSavingsMsg'
Learn More

Mabuterol, TRC

Product Code. 30009724
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009724 100 mg 100mg
17932485 10 mg 10mg
17980995 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009724 Supplier Toronto Research Chemicals Supplier No. TRCM104000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56341-08-3
Molecular Formula C13 H18 Cl F3 N2 O
Molecular Weight (g/mol) 310.74
Synonym Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)-, 4-Amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)benzenemethanol, (±)-Mabuterol, Ambuterol, Mabuterol, PB 868Cl
IUPAC Name 1-[4-amino-3-chloro-5-(trifluoromethyl)phenyl]-2-(tert-butylamino)ethanol
SMILES CC(C)(C)NCC(O)c1cc(Cl)c(N)c(c1)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H18ClF3N2O/c1-12(2,3)19-6-10(20)7-4-8(13(15,16)17)11(18)9(14)5-7/h4-5,10,19-20H,6,18H2,1-3H3
Formula Weight 310.106
Percent Purity >95%
Chemical Name or Material Mabuterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.