accessibility menu, dialog, popup

UserName

CH3CH3H3COONOOHOOH

CAS RN 56074-20-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl](carboxymethyl)amino}acetic acid
Synonyms: [(tert-butoxycarbonyl)(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C9H15NO6
InChi Key: QPGIJQUTGABQLQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N(CC(O)=O)CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)