missing translation for 'onlineSavingsMsg'
Learn More

Boc-Iminodiacetic Acid, TRC

Product Code. 30335186
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335186 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335186 Supplier Toronto Research Chemicals Supplier No. TRCB64846350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-Iminodiacetic Acid
CAS 56074-20-5
Molecular Formula C9H15NO6
Quantity 50 mg
Synonym 2-[Carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic Acid, 2-[[(tert-Butoxy)carbonyl](carboxymethyl)amino]acetic Acid, 3-(tert-Butoxycarbonyl)-3-azapentanedioic Acid, N-(tert-Butoxycarbonyl)iminodiacetic Acid, N-Boc-iminodiacetic Acid, [(tert-Butoxycarbonyl)(carboxymethyl)amino]acetic Acid
InChI Formula InChI=1S/C9H15NO6/c1-9(2,3)16-8(15)10(4-6(11)12)5-7(13)14/h4-5H2,1-3H3,(H,11,12)(H,13,14)
SMILES CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O
IUPAC Name 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
Molecular Weight (g/mol) 233.21
Formula Weight 233.0899
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.