accessibility menu, dialog, popup

UserName

CH3NNH2

CAS RN 53485-07-7

IUPAC Name: N-(3-aminopropyl)-N-methylaniline
Synonyms: N-(3-aminopropyl)-N-methylaniline
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: WGHBKYWUOOANEX-UHFFFAOYSA-N
SMILES: CN(CCCN)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results