missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)-N-methylaniline, TRC

Product Code. 30323559
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323559 Supplier Toronto Research Chemicals Supplier No. TRCA6293281G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Aminopropyl)-N-methylaniline
CAS 53485-07-7
Molecular Formula C10H16N2
Quantity 1 g
Synonym N1-Methyl-N1-phenyl-1,3-propanediamine, 3-(Methylphenylamino)propylamine, 3-(N-Methylanilino)propylamine, N-(3-Aminopropyl)-N-methylaniline, N-Methyl-N-phenyl-1,3-propanediamine, N1-Methyl-N1-phenylpropan-1,3-diamine, N1-Methyl-N1-phenylpropane-1,3-diamine, N1-Methyl-N1-phenylpropane-1,3-diamine
InChI Formula InChI=1S/C10H16N2/c1-12(9-5-8-11)10-6-3-2-4-7-10/h2-4,6-7H,5,8-9,11H2,1H3
SMILES CN(CCCN)c1ccccc1
IUPAC Name N'-methyl-N'-phenylpropane-1,3-diamine
Molecular Weight (g/mol) 164.25
Formula Weight 164.1313
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.