accessibility menu, dialog, popup

UserName

CAS RN 524-95-8

IUPAC Name: 2-aminoethyl diphenylborinate
Synonyms: 2-aminoethoxydiphenyl borate
Molecular Weight (g/mol): 225.1
Molecular Formula: C14H16BNO
InChi Key: BLZVCIGGICSWIG-UHFFFAOYSA-N
SMILES: NCCOB(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results