CAS RN 51534-66-8
IUPAC Name:
methyl (2S)-2-(methylsulfanyl)butanoate
Synonyms:
methyl (2S)-2-(methylsulfanyl)butanoate
Molecular Weight (g/mol):
148.22
Molecular Formula:
C6H12O2S
InChi Key:
PBYSEUNXLXTEAN-YFKPBYRVSA-N
SMILES:
CC[C@H](SC)C(=O)OC
1
–
1
of
1
results
1
Methyl 2-(methylthio)butyrate, 98%, Thermo Scientific™
CAS: 51534-66-8 Molecular Formula: C6H12O2S Molecular Weight (g/mol): 148.22 MDL Number: MFCD00013393 InChI Key: PBYSEUNXLXTEAN-UHFFFAOYSA-N PubChem CID: 560572 IUPAC Name: methyl 2-methylsulfanylbutanoate SMILES: CCC(C(=O)OC)SC