Learn More
Methyl 2-(methylthio)butyrate, 98%, Thermo Scientific™
Product Code. 11435986
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435986 25 g 25g
11425986 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11435986 Supplier Thermo Scientific Alfa Aesar Supplier No. B22303.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51534-66-8
Molecular Formula C6H12O2S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00013393
InChI Key PBYSEUNXLXTEAN-UHFFFAOYSA-N
PubChem CID 560572
IUPAC Name methyl 2-methylsulfanylbutanoate
SMILES CCC(C(=O)OC)SC
Specifications
Density 1.028
Boiling Point 103°C to 104°C (35 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.461
Quantity 25 g
Beilstein 1925141
Synonym methyl 2-methylthio butyrate, methyl 2-methylsulfanyl butanoate, butanoic acid,2-methylthio-, methyl ester, methyl 2-methylthio-butyrate, methyl alpha-methylsulfanyl butyrate, acmc-20akj9, 2-methylthio-butanoic acid methyl ester, butanoic acid, 2-methylthio-, methyl ester, +/--butanoic acid, 2-methylthio-, methyl ester, butanoic acid, 2-methylthio-, methyl ester, +/-
Formula Weight 148.23
Percent Purity 98%
Chemical Name or Material Methyl 2-(methylthio)butyrate