accessibility menu, dialog, popup

UserName

CAS RN 51173-04-7

IUPAC Name: 5-fluoro-2-methoxypyridine
Synonyms: 5-fluoro-2-methoxypyridine
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: GYGABXVZIHADCT-UHFFFAOYSA-N
SMILES: COC1=NC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results