missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxypyridine, 97%, Thermo Scientific™

Product Code. 15474017
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474017 25 g 25g
15454017 1 g 1g
15464017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15474017 Supplier Thermo Scientific Alfa Aesar Supplier No. H30991.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51173-04-7
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD03095252
InChI Key GYGABXVZIHADCT-UHFFFAOYSA-N
Synonym 2-methoxy-5-fluoropyridine, pyridine, 5-fluoro-2-methoxy, 5-fluoro-2-methoxy-pyridine, pubchem11248, acmc-1b1gh, 3-fluoro-6-methoxypyridine, 5-fluoro-2-methoxy pyridine, ksc497o4b, 5-fluoropyridin-2-yl methyl ether, abbypharma ap-14-5659
PubChem CID 2783175
IUPAC Name 5-fluoro-2-methoxypyridine
SMILES COC1=NC=C(C=C1)F

Specifications

Quantity 25 g
UN Number UN1993
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.