CAS RN 504-96-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Neophytadiene (80%), TRC
CAS: 504-96-1 Molecular Formula: C20 H38 Molecular Weight (g/mol): 278.52 Synonym: 7,11,15-Trimethyl-3-methylene-1-hexadecene,2-(4,8,12-Trimethyltridecyl)-1,3-butadiene,2-(4,8,12-Trimethyltridecyl)buta-1,3-diene,3-Methylene-7,11,15-trimethylhexadec-1-ene; IUPAC Name: 7,11,15-trimethyl-3-methylidenehexadec-1-ene SMILES: CC(C)CCCC(C)CCCC(C)CCCC(=C)C=C